Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride, 99%, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
7447-41-8
ClLi
42.38
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
7447-41-8
ClLi
42.38
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
lithium chloride,lithium chloride licl,lithiumchloride,licl,chlorure de lithium,chlorku litu,chlorolithium,lithiumchlorid,chlorku litu polish,luthium chloride
433294
CHEBI:48607
lithium;chloride
[Li+].[Cl-]
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15,5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols,ether,pyridine,,nitrobenzene and acetone
Formula Weight 42.38
Percent Purity 99%
Grade Extra Pure
Density 2.0600g/mL
Insoluble Matter 0.01% max. (in acid)
Packaging Plastic bottle
Solubility Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone
Specific Gravity 2.06
Boiling Point 1382.0°C
Melting Point 605.0°C
Quantity 5kg
Chemical Name or Material Lithium chloride
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 231-212-3