Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium cobalt(III) oxide, 97%, Thermo Scientific Chemicals

Catalog Number p-4407412
Click to view available options
Quantity:
50 g
250 g
1 kg
12190-79-3
CoLiO2
97.871
MFCD00049786
BFZPBUKRYWOWDV-UHFFFAOYSA-N
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
23670860
lithium;oxido(oxo)cobalt
[Li+].[O-][Co]=O
12190-79-3
CoLiO2
97.871
MFCD00049786
BFZPBUKRYWOWDV-UHFFFAOYSA-N
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
23670860
lithium;oxido(oxo)cobalt
[Li+].[O-][Co]=O

Lithium cobalt(III) oxide is a critical substance in the quest for high performance, advanced batteries. It is used as a lithium ion cathode material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue
Physical Form Powder
Quantity 50 g
Assay Percent Range 97%
Solubility Information Insoluble in water.
Formula Weight 97.88
Odor Odorless
Sensitivity Hygroscopic
Appearance Dark blue powder
Chemical Name or Material Lithium cobalt(III) oxide
Hazard Category H350-H361f
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 235-362-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit