Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium deuteride, 98%(Isotopic), Alfa Aesar™

Used in thermonuclear Fusion

Manufacturer:  Alfa Aesar 4410004

Quantity 2g

 View more versions of this product

Catalog No. AA4410004


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13587-16-1
98% (Isotopic)
8.95
Lithium deuteride
Powder
DLi
MFCD00011091

Specifications

13587-16-1
98% (Isotopic)
Gray powder
MFCD00011091
200°C (392°F)
Gray
Lithium deuteride
Powder
DLi
8.95
0.906g/cm3 at 20°C
Odorless
2g

Safety and Handling

SDS & Safety Documents

H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 237-018-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit