Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium deuteride, 99% atom D, for NMR, ACROS Organics™
Click to view available options
Quantity:
2g
10g
Packaging:
Plastic bottle
Description
For NMR
Specifications
Deuterium (D) | 98.5% min. |
Density | 0.8200g/mL |
Packaging | Plastic bottle |
Quantity | 10g |
Assay Percent Range | 99 atom% D |
Linear Formula | LiD |
Formula Weight | 8.96 |
Physical Form | Crystalline |
Grade | NMR |
Chemical Name or Material | Lithium deuteride, 99 atom % D |
Safety and Handling
GHS H Statement:
In contact with water releases flammable gas.
GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 237-018-5