Alfa Aesar™ Lithium dimethylamide, 10% w/v in hexane

Manufacturer: Alfa Aesar™ 45491AK

Quantity 250mL

 View more versions of this product

Catalog No. AA45491AK

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 3585-33-9
    Molecular Formula C2H6LiN
    Linear Formula LiN(CH3)2
    Formula Weight 51.02
    MDL Number MFCD00008295
    UN Number UN3394
    Chemical Name or Material Lithium dimethylamide

    Specifications

    Formula Weight 51.02
    Health Hazard 2 GHS H Statement
    H250-H260-H252-H304-H361-H373-H314-H318-H336
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities; may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P301+P330+P331-P315-P403
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.
    Store in a well-ventilated place.
    Molecular Formula C2H6LiN
    Linear Formula LiN(CH3)2
    UN Number UN3394
    Concentration 10% w/v in hexane
    Color White
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    MDL Number MFCD00008295
    CAS 3585-33-9
    Sensitivity Air & Moisture Sensitive
    Physical Form Slurry
    Quantity 250mL
    Odor Petrol-like
    Chemical Name or Material Lithium dimethylamide

    GHS H Statement
    H250-H260-H252-H304-H361-H373-H314-H318-H336
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities; may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280-P305+P351+P338-P301+P330+P331-P315-P403
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit