Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lithium Dodecylsulfate, ≥98%, Thermo Scientific™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Plastic bottle
2044-56-6
C12H25LiO4S
272.328
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium dodecyl sulfate,lithium dodecylsulfate,sulfuric acid, monododecyl ester, lithium salt,lauryl sulfate lithium salt,lithium lauryl sulfate,dodecyl lithium sulfate,lds,lithium 1+ ion dodecyl sulfate,sulfuric acid, monododecyl ester, lithium salt 1:1,lithium dodecyl sulfate, ultra pure
2735071
lithium;dodecyl sulfate
[Li+].CCCCCCCCCCCCOS(=O)(=O)[O-]
2044-56-6
C12H25LiO4S
272.328
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium dodecyl sulfate,lithium dodecylsulfate,sulfuric acid, monododecyl ester, lithium salt,lauryl sulfate lithium salt,lithium lauryl sulfate,dodecyl lithium sulfate,lds,lithium 1+ ion dodecyl sulfate,sulfuric acid, monododecyl ester, lithium salt 1:1,lithium dodecyl sulfate, ultra pure
2735071
lithium;dodecyl sulfate
[Li+].CCCCCCCCCCCCOS(=O)(=O)[O-]
Linear Formula CH3(CH2)11OSO3Li
UN Number UN1325
Formula Weight 272.33
Physical Form Powder
Percent Purity ≥98%
Packaging Plastic bottle
Quantity 25g
Chemical Name or Material Lithium Dodecylsulfate

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302+H332-H315-H319-H335

DOTInformation : Hazard Class: 4.1

EINECSNumber : 218-058-2

TSCA : Yes

Recommended Storage : Ambient temperatures