Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium dodecylsulfate, 99+%

Manufacturer:  Alfa Aesar™ 3932809

Quantity 10g

 View more versions of this product

Catalog No. AA3932809


  Check Availability
Add to cart

Specifications

Description

  • Detergent for solubilizing proteins for electrophoresis which exhibits greater solubility than SDS at lower temperatures
Description & Specifications

Chemical Identifiers

2044-56-6
MFCD00007467
Lithium dodecylsulfate
272.33
UN1325

Specifications

2044-56-6
White
Powder
MFCD00007467
Lithium dodecylsulfate
10g
Odorless
272.33
UN1325
≥99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H228-H335-H315
Causes serious eye damage.
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit