Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium fluoride, 99.98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
7789-24-4
FLi
25.93
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif
224478
lithium;fluoride
[Li+].[F-]
7789-24-4
FLi
25.93
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride,fluorolithium,lithium monofluoride,trilithium trifluoride,lithium fluoride li3f3,lithium fluorure,ntl 50,lithium fluoride 6lif,lithium fluoride 7lif
224478
lithium;fluoride
[Li+].[F-]
Assay Percent Range 99.98%
Linear Formula LiF
Fieser 01,933
Merck Index 15, 5587
Solubility Information Solubility in water: 0.29g/100mL (20°C). Other solubilities: soluble in acids,insoluble in alcohol
Formula Weight 25.93
Physical Form Powder
Percent Purity 99.98%
Grade Extra Pure
Density 2.6300g/mL
Packaging Plastic bottle
Solubility Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol
Specific Gravity 2.63
Total Trace Metal Impurities 0.02% max.
Boiling Point 1681.0°C
Color White
Melting Point 845.0°C
Quantity 25g
Chemical Name or Material Lithium fluoride
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist

GHS Signal Word: Danger

EINECSNumber : 232-152-