Lithium fluoride, White powder, 99.99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1446318

Quantity 50g

 View more versions of this product

Catalog No. AA1446318

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7789-24-4
    Physical Form Powder
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula FLi
    Formula Weight 25.94
    MDL Number MFCD00011090
    Chemical Name or Material Lithium fluoride

    Specifications

    Physical Form Powder
    Formula Weight 25.94
    Quantity 50g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    Health Hazard 2 Keep away from living quarters. Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    EINECS Number 232-152-0
    Odor Odorless
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula FLi
    MDL Number MFCD00011090
    Health Hazard 1 H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    CAS 7789-24-4
    Chemical Name or Material Lithium fluoride

    Keep away from living quarters. Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    H301-H315-H319-H335: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit