Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hexafluorophosphate, 98%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
21324-40-3
F6LiP
151.91
MFCD00011096
AXPLOJNSKRXQPA-UHFFFAOYSA-N
lithium hexafluorophosphate,lithium hexafluorophosphate v,phosphate 1-, hexafluoro-, lithium,phosphate 1-, hexafluoro-, lithium 1:1,lithiumhexafluorophosphate,lithium hexafluorophosphate 1-,lipf6,lithium hexafluoroposphate,acmc-209fj7,ksc492g4h
23688915
lithium;hexafluorophosphate
[Li+].F[P-](F)(F)(F)(F)F
21324-40-3
F6LiP
151.91
MFCD00011096
AXPLOJNSKRXQPA-UHFFFAOYSA-N
lithium hexafluorophosphate,lithium hexafluorophosphate v,phosphate 1-, hexafluoro-, lithium,phosphate 1-, hexafluoro-, lithium 1:1,lithiumhexafluorophosphate,lithium hexafluorophosphate 1-,lipf6,lithium hexafluoroposphate,acmc-209fj7,ksc492g4h
23688915
lithium;hexafluorophosphate
[Li+].F[P-](F)(F)(F)(F)F
Assay Percent Range 97.5% min. (ex Li)
Linear Formula LiPF6
Solubility Information Solubility in water: hydrolysis
Formula Weight 151.91
Physical Form Fuming Powder
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Solubility Solubility in water: hydrolysis.
Color White
Quantity 5g
Chemical Name or Material Lithium hexafluorophosphate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 244-334-7