Alfa Aesar™ Lithium hexafluorosilicate

Manufacturer: Alfa Aesar™ 3933206

Quantity 5g

 View more versions of this product

Catalog No. AA3933206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17347-95-4
    Formula Weight 155.96
    MDL Number MFCD00049473
    UN Number UN2856
    Chemical Name or Material Lithium hexafluorosilicate

    Specifications

    Formula Weight 155.96
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Physical Form Powder
    CAS 17347-95-4
    UN Number UN2856
    Quantity 5g
    TSCA TSCA
    Odor Odorless
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROSILICATES, N.O.S.
    MDL Number MFCD00049473
    Chemical Name or Material Lithium hexafluorosilicate

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit