Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydride, 97%, pure, powder (<100 µm), in resealable cans, Acros Organics

Click to view available options
Quantity:
500g
7580-67-8
HLi
7.95
MFCD00011074
SRTHRWZAMDZJOS-UHFFFAOYSA-N
lithium hydride,lithiumhydride,lih,lithium hydride anion,hydride lithium,hydridolithate 1-,lih-,ksc491a4n
62714
lithium;hydride
[H-].[Li+]
7580-67-8
HLi
7.95
MFCD00011074
SRTHRWZAMDZJOS-UHFFFAOYSA-N
lithium hydride,lithiumhydride,lih,lithium hydride anion,hydride lithium,hydridolithate 1-,lih-,ksc491a4n
62714
lithium;hydride
[H-].[Li+]
Reduction reagent
Assay Percent Range 96% min
Linear Formula LiH
Fieser 04,304; 13,165
Merck Index 15, 5589
Solubility Information Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene,soluble in ether
Formula Weight 7.95
Physical Form Powder
Percent Purity 97%
Grade Pure
Density 0.8200g/mL
Packaging Plastic bag in resealable tin can
Solubility Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene, soluble in ether
Specific Gravity 0.82
Color White to Gray
Melting Point 680.0°C
Quantity 500g
Chemical Name or Material Lithium hydride, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Handle under inert gas.
Protec

GHS Signal Word: Danger

EINECSNumber : 231-484-3