Learn More
Lithium hydroxide, anhydrous, 99.995% (metals basis), Thermo Scientific Chemicals
Description
It is used as a heat transfer medium, as a storage-battery electrolyte and also used for the production of lithium greases. It is also used in ceramics, in some portland cement formulations, in the absoption of carbondioxide from sealed enviornments such as submarines, spacecrafts and breathing apparatus. It is used in esterification reactions, as stabilizer in photographic developments and as a coolant in pressurized water reactors for corrosion control.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 924°C |
Melting Point | 462°C |
Color | White |
pH | 14 |
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN2680 |
Merck Index | 14,5534 |
Solubility Information | Soluble in water. Slightly soluble in ethanol and methanol |
Formula Weight | 23.95 |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335-H336-H373 |
Hazard Statement | GHS H Statement H301-H331-H314-H318 Toxic if swallowed. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE |
EINECSNumber | 215-183-4 |
RTECSNumber | OJ6307070 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.