Lithium hydroxide monohydrate, 56.5% min, Alfa Aesar™

Manufacturer: Alfa Aesar 1340918

Quantity 50g

 View more versions of this product

Catalog No. AA1340918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310-66-3
    Physical Form Powder
    Assay Percent Range 56.5% min
    Molecular Formula LiOH·H2O
    Formula Weight 41.96; 23.95 (anhydrous)
    MDL Number MFCD00149772
    Chemical Name or Material Lithium hydroxide monohydrate

    Specifications

    Formula Weight 41.96; 23.95 (anhydrous)
    Quantity 50g
    EINECS Number 215-183-4
    Physical Form Powder
    Density 1.51g/mL
    Molecular Formula LiOH·H2O
    CAS 1310-66-3
    Assay Percent Range 56.5% min
    MDL Number MFCD00149772
    Chemical Name or Material Lithium hydroxide monohydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit