Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Hydroxide, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

1310-66-3
H3LiO2
41.962
GLXDVVHUTZTUQK-UHFFFAOYSA-M
168937
lithium;hydroxide;hydrate
[Li+].O.[OH-]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 41.96
Packaging Poly Bottle
Specific Gravity 1L = 1.51kg
Quantity 1kg
Chemical Name or Material Lithium Hydroxide, Monohydrate
Grade Baker Analyzed™ ACS Reagent