Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
1310-66-3
H3LiO2
41.962
GLXDVVHUTZTUQK-UHFFFAOYSA-M
lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water
168937
lithium;hydroxide;hydrate
[Li+].O.[OH-]
1310-66-3
H3LiO2
41.962
GLXDVVHUTZTUQK-UHFFFAOYSA-M
lithium hydroxide monohydrate,lithium hydroxide hydrate,lithium hydroxido,hydroxyde de lithium,unii-g51xlp968g,lithium hydroxide, monohydrate,lithiumhydrate,lioh-hydrate,lioh.hydrate,lioh water
168937
lithium;hydroxide;hydrate
[Li+].O.[OH-]
Aluminum (Al) 300ppm max.
Linear Formula LiOH·H2O
Formula Weight 41.95
Physical Form Crystalline Powder
Percent Purity ≥54% (LiOH)
Grade Extra Pure
Carbon Dioxide 3500ppm max.
Chloride 500ppm max.
Insoluble in Water 0.1% max.
Iron (Fe) 10ppm max.
Packaging Plastic drum
Sodium and Potassium 3000ppm max.
Solubility Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol
Sulfate 500ppm max.
Boiling Point 920.0°C
Color Cream to White or Yellow
Melting Point 462.0°C
Quantity 5kg
Chemical Name or Material Lithium hydroxide monohydrate
Show More Show Less