Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium ingot, 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 00658KI

Quantity 4 pieces

 View more versions of this product

Catalog No. AA00658KI


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7439-93-2
MFCD00134051
UN1415
Lithium ingot, 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long

Specifications

≈116g/pc
7439-93-2
MFCD00134051
Odorless
Silver-white
99.9%, (metals basis)
4 pieces
Pieces
UN1415
Vacuum can
Lithium ingot, 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long

Safety and Handling

SDS & Safety Documents

GHS H Statement
H260-H314
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM

RTECSNumber : OJ5540000

TSCA : TSCA

storageNote1 :

  • In vacuum can with mineral oil coating

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit