Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium iodide, 99%, pure, anhydrous, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
10377-51-2
ILi
133.85
MFCD00011092
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide
66321
lithium;iodide
[Li+].[I-]
10377-51-2
ILi
133.85
MFCD00011092
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide
66321
lithium;iodide
[Li+].[I-]
Assay Percent Range 98.5% min. (on anhydrous substance) (Argentometry)
Linear Formula LiI
Fieser 01,615; 04,304; 05,410; 07,208; 09,283; 10,245; 11,300; 12,282; 15,185; 16,197; 17,85
Merck Index 15, 5591
Solubility Information Solubility in water: 165g/100 mL (20°C). Other solubilities: soluble in alcohol,freely soluble in acetone
Formula Weight 133.85
Physical Form Powder, Crystals and/or Granules
Percent Purity 99%
Grade Pure
Density 3.49
Packaging Glass bottle
Solubility Solubility in water: 165 g/100 mL (20°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Water 1% max. (K.F.)
Boiling Point 1171.0°C
Color Undesignated
Melting Point 446.0°C
Quantity 50g
Chemical Name or Material Lithium iodide, 99%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Warning

EINECSNumber : 233-822-5

TSCA : TSCA