Alfa Aesar™ Lithium iodide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1360014

Quantity 25g

 View more versions of this product

Catalog No. AA1360014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10377-51-2
    Formula Weight 133.84
    MDL Number MFCD00011092
    Merck Index 14,5535
    Chemical Name or Material Lithium iodide, Ultra dry

    Specifications

    Formula Weight 133.84
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Packaging Ampule
    Odor Odorless
    Merck Index 14,5535
    Storage Note 1
    • Ampouled under argon
    Density 4.076g/mL
    Physical Form Beads
    MDL Number MFCD00011092
    CAS 10377-51-2
    Boiling Point 1170° to 1190°C
    Color White to off-white
    Mesh Size −10 Mesh
    Refractive Index 1.955
    Sensitivity Light Sensitive & Hygroscopic
    Quantity 25g
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 449°C
    Health Hazard 1 Warning
    Chemical Name or Material Lithium iodide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit