Alfa Aesar™ Lithium isobutyrate, 98+%

Manufacturer: Alfa Aesar™ 3937222

Quantity 100g

 View more versions of this product

Catalog No. AA3937222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25179-23-1
    Formula Weight 94.03
    MDL Number MFCD00210622
    Chemical Name or Material Lithium isobutyrate

    Specifications

    Physical Form Powder/Lump
    Formula Weight 94.03
    Quantity 100g
    MDL Number MFCD00210622
    CAS 25179-23-1
    Sensitivity Moisture Sensitive
    Chemical Name or Material Lithium isobutyrate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit