Alfa Aesar™ Lithium isopropoxide, 94%

Manufacturer: Alfa Aesar™ 1635203

Quantity 1g

 View more versions of this product

Catalog No. AA1635203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2388-10-5
    Linear Formula (CH3)2CHOLi
    Formula Weight 66.03
    MDL Number MFCD00048286
    UN Number UN3206
    Beilstein 3618614
    Chemical Name or Material Lithium isopropoxide

    Specifications

    Formula Weight 66.03
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place. Store in a closed container.
    Physical Form Powder
    Linear Formula (CH3)2CHOLi
    UN Number UN3206
    Beilstein 3618614
    Health Hazard 2 GHS H Statement
    H251-H314
    Self-heating: may catch fire.
    Causes severe skin burns and eye damage.
    CAS 2388-10-5
    Quantity 1g
    Health Hazard 1 Danger
    Color White
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00048286
    Chemical Name or Material Lithium isopropoxide
    Assay 94%

    GHS H Statement
    H251-H314
    Self-heating: may catch fire.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place. Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit