Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium isopropoxide, 94%

Manufacturer:  Alfa Aesar™ 1635206

Quantity 5g

 View more versions of this product

Catalog No. AA1635206


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2388-10-5
66.03
UN3206
Lithium isopropoxide
(CH3)2CHOLi
MFCD00048286
3618614

Specifications

2388-10-5
White
Powder
66.03
UN3206
Lithium isopropoxide
5g
Air & Moisture Sensitive
(CH3)2CHOLi
MFCD00048286
3618614
94%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit