Alfa Aesar™ Lithium L-lactate, 97%

Manufacturer: Alfa Aesar™ A118180B

Quantity 1000g

 View more versions of this product

Catalog No. AAA118180B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27848-80-2
    Molecular Formula C3H5LiO3
    Formula Weight 96.01
    MDL Number MFCD00065399
    Beilstein 3568332
    Synonym L-Lactic acid lithium salt
    Chemical Name or Material Lithium L-lactate

    Specifications

    Formula Weight 96.01
    Molecular Formula C3H5LiO3
    TSCA TSCA
    MDL Number MFCD00065399
    Beilstein 3568332
    Synonym L-Lactic acid lithium salt
    Melting Point 253° to 255°C
    Quantity 1000g
    Optical Rotation −14° (c=1 in water)
    CAS 27848-80-2
    RTECS Number OJ6311000
    Chemical Name or Material Lithium L-lactate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit