Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium metaborate, 99%, for analysis, Thermo Scientific Chemicals

Catalog Number AC365711000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
lithium;oxido(oxo)borane
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
lithium;oxido(oxo)borane
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 845°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula LiBO2
Solubility Information Solubility in water: 25.7g/L (20°C)
Formula Weight 49.75
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Lithium metaborate, 99%
Show More Show Less
EINECSNumber 236-631-5
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit