Lithium metaborate, anhydrous, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 12591A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12591A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13453-69-5
    Physical Form Powder
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula BLiO2
    Formula Weight 49.75
    MDL Number MFCD00011089
    Chemical Name or Material Lithium metaborate, anhydrous

    Specifications

    Formula Weight 49.75
    Quantity 1kg
    EINECS Number 236-631-5
    Physical Form Powder
    Density 1.397g/mL
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula BLiO2
    MDL Number MFCD00011089
    CAS 13453-69-5
    Chemical Name or Material Lithium metaborate, anhydrous

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit