Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium metaborate, anhydrous, tech., Thermo Scientific Chemicals

Catalog Number p-7049982
Click to view available options
Quantity:
100 g
500 g
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

Lithium Metaborate is commonly used as a fused compound or a component for an assay. It has been used for the analysis of wild rice for silicon, calcium, magnesium and potassium. Lithium metaborate was used to fuse with solid samples for decomposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 845°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula LiBO2
Solubility Information Soluble in water (25.7mg/ml at 20°C).
IUPAC Name tetraboron heptahydrate dilithium
Formula Weight 49.75
Grade Technical
Odor Odorless
Sensitivity Hygroscopic
Density 1.397 g/mL
Chemical Name or Material Lithium metaborate, Anhydrous
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 236-631-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit