Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium Metaborate (100%), Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™
Click to view available options
Quantity:
1kg
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Pure Grade Fusion Flux
- 1 kg
- Lithium Metaborate (100%)
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Concentration or Composition (by Analyte or Components) | Various |
Physical Form | Crystalline |
Packaging | Triple-leached LDPE bottle |
Certificate of Analysis | With Certificate of Analysis. |
Quantity | 1kg |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Chemical Name or Material | Lithium Metaborate |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only