Alfa Aesar™ Lithium metagallate, 99%

Manufacturer: Alfa Aesar™ 3937014

Quantity 25g

 View more versions of this product

Catalog No. AA3937014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 944251-30-3
    Linear Formula LiGaO2
    Formula Weight 108.66
    MDL Number MFCD00210623
    Synonym Lithium gallate(III); Gallium(III) lithium oxide
    Chemical Name or Material Lithium metagallate

    Specifications

    Formula Weight 108.66
    Physical Form Powder
    Synonym Lithium gallate(III); Gallium(III) lithium oxide
    Color White
    Quantity 25g
    Linear Formula LiGaO2
    MDL Number MFCD00210623
    CAS 944251-30-3
    Melting Point >400°C
    Chemical Name or Material Lithium metagallate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit