Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol

Click to view available options
Quantity:
25mL
100mL
86745-52-0
C12H30LiNbO6
370.212
MFCD00210625
AVEKRHJBCCYCED-UHFFFAOYSA-N
lithium niobium ethoxide metals basis w/v in ethanol,lithium niobium ethoxide,lithium niobium 5+ ethoxide 1/1/6,niobium 5+ lithium 1+ hexakis ethoxide
14693679
lithium;ethanolate;niobium(5+)
[Li+].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
86745-52-0
C12H30LiNbO6
370.212
MFCD00210625
AVEKRHJBCCYCED-UHFFFAOYSA-N
lithium niobium ethoxide metals basis w/v in ethanol,lithium niobium ethoxide,lithium niobium 5+ ethoxide 1/1/6,niobium 5+ lithium 1+ hexakis ethoxide
14693679
lithium;ethanolate;niobium(5+)
[Li+].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
Quantity 100mL
Sensitivity Moisture sensitive
Flash Point 17°C (63°F)
Odor Alcohol-like
Color Yellow
UN Number UN1170
Formula Weight 370.21
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Lithium niobium ethoxide
Assay (metals basis)
Show More Show Less

GHS H Statement
H225-H315-H319
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen