Alfa Aesar™ Lithium niobium tert-butoxide, 99+% (metals basis), 5% w/v in tert-butanol

Manufacturer: Alfa Aesar™ 39157AC

Quantity 25mL

 View more versions of this product

Catalog No. AA39157AC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 21864-27-7
    Linear Formula LiNb(OC4H9)6
    Formula Weight 538.54
    MDL Number MFCD00210624
    UN Number UN1120
    Chemical Name or Material Lithium niobium tert-butoxide

    Specifications

    Formula Weight 538.54
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00210624
    Quantity 25mL
    UN Number UN1120
    Health Hazard 2 GHS H Statement
    H225-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Alcohol-like
    Sensitivity Moisture Sensitive
    Concentration 5% w/v in tert-butanol
    Linear Formula LiNb(OC4H9)6
    Color Yellow to brown
    Flash Point 11° (52°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS
    CAS 21864-27-7
    Health Hazard 1 Danger
    Chemical Name or Material Lithium niobium tert-butoxide
    Assay ≥99%, (metals basis)

    GHS H Statement
    H225-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit