Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium nitrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
7790-69-4
LiNO3
68.95
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
lithium;nitrate
[Li+].[N+](=O)([O-])[O-]
7790-69-4
LiNO3
68.95
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate,nitric acid, lithium salt,lithium nitrate, anhydrous,lithium 1+ ion nitronate,ksc492i7j,lithium nitrate 3-hydrate,lithium 1+ ion nitrate,lithium nitrate, anhydrous 25g,lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
lithium;nitrate
[Li+].[N+](=O)([O-])[O-]
Alkalinity 0.05% max.
Assay Percent Range 99+%
Linear Formula LiNO3
Merck Index 15, 5592
Solubility Information Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol
Formula Weight 68.95
Physical Form Adhering Crystalline Powder
Percent Purity 99+%
Grade Extra Pure
Chlorine (Cl) 100ppm max.
Density 2.3800g/mL
Fe2O3 50ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol
Specific Gravity 2.38
Sulfate 500ppm max.
Water 1% max.
Boiling Point 600.0°C
Color White
Melting Point 251.0°C
Quantity 250g
Chemical Name or Material Lithium nitrate
Show More Show Less

GHS H Statement
Harmful if swallowed.
May intensify fire; oxidizer.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Warning

EINECSNumber : 232-218-9