Lithium nitrate, anhydrous, 99%, Alfa Aesar™

Used in Ceramics, Pyrotechnics, Salt Baths

Manufacturer: Alfa Aesar 13405A1

Quantity 1kg

 View more versions of this product

Catalog No. AA13405A1

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7790-69-4
    Physical Form Crystals
    Assay Percent Range 99%
    Molecular Formula LiNO3
    Formula Weight 68.95
    MDL Number MFCD00011094
    Chemical Name or Material Lithium nitrate, anhydrous

    Specifications

    Physical Form Crystals
    Formula Weight 68.95
    Quantity 1kg
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 232-218-9
    Appearance White crystalline
    MDL Number MFCD00011094
    CAS 7790-69-4
    Odor Odorless
    Assay Percent Range 99%
    Molecular Formula LiNO3
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Lithium nitrate, anhydrous

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit