Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Nitrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7790-69-4
LiNO3
68.944
IIPYXGDZVMZOAP-UHFFFAOYSA-N
10129889
CHEBI:63315

High quality chemicals for laboratory and specialized industrial use

CAS 7790-69-4
Molecular Formula LiNO3
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES [Li+].[N+](=O)([O-])[O-]
IUPAC Name lithium;nitrate
Molecular Weight (g/mol) 68.944
PubChem CID 10129889
ChEBI CHEBI:63315
Formula Weight 68.94
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.38kg
Quantity 2.5kg
Chemical Name or Material Lithium Nitrate
Grade BAKER ANALYZED™ Reagent
Show More Show Less