Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium octanoate, 99%

Click to view available options
Quantity:
25g
100g
500g
16577-52-9
C8H15LiO2
150.146
MFCD00210629
BTAUEIDLAAYHSL-UHFFFAOYSA-M
lithium octanoate,unii-9nq46xlp8b,octanoic acid, lithium salt,9nq46xlp8b,octanoic acid, lithiumsalt 1:1,lithium caprylate,octanoic acid, lithium salt 1:1,acmc-20akmi,lithium 1+ octanoate,lithium 1+ ion octanoate
23673717
lithium;octanoate
[Li+].CCCCCCCC(=O)[O-]
16577-52-9
C8H15LiO2
150.146
MFCD00210629
BTAUEIDLAAYHSL-UHFFFAOYSA-M
lithium octanoate,unii-9nq46xlp8b,octanoic acid, lithium salt,9nq46xlp8b,octanoic acid, lithiumsalt 1:1,lithium caprylate,octanoic acid, lithium salt 1:1,acmc-20akmi,lithium 1+ octanoate,lithium 1+ ion octanoate
23673717
lithium;octanoate
[Li+].CCCCCCCC(=O)[O-]
Quantity 100g
Formula Weight 150.14
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Lithium octanoate

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 240-637-3

TSCA : Yes

Recommended Storage : Ambient temperatures