Alfa Aesar™ Lithium perchlorate, anhydrous, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4422630

Quantity 250g

 View more versions of this product

Catalog No. AA4422630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7791-03-9
    Molecular Formula LiClO4
    Formula Weight 106.4
    MDL Number MFCD00011079
    UN Number UN1481
    Merck Index 14,5539
    Chemical Name or Material Lithium perchlorate, Anhydrous

    Specifications

    Formula Weight 106.4
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 7791-03-9
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.43g/mL
    Physical Form Lumps
    Boiling Point 400°C (dec.)
    Melting Point 236°C
    Quantity 250g
    Molecular Formula LiClO4
    Merck Index 14,5539
    Health Hazard 1 Danger
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    Sensitivity Hygroscopic
    MDL Number MFCD00011079
    UN Number UN1481
    Color White
    Chemical Name or Material Lithium perchlorate, Anhydrous
    Assay 99.99%, (metals basis)

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit