Lithium perchlorate, anhydrous, 99%, Alfa Aesar™

For Lithium Battery

Manufacturer: Alfa Aesar 4422518

Quantity 50g

 View more versions of this product

Catalog No. AA4422518

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7791-03-9
    Physical Form Crystals
    Assay Percent Range 99%
    Molecular Formula ClLiO4
    Formula Weight 106.4
    MDL Number MFCD00011079
    Chemical Name or Material Lithium perchlorate, anhydrous

    Specifications

    Physical Form Crystals
    Formula Weight 106.4
    Quantity 50g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: II
    EINECS Number 232-237-2
    Odor Odorless
    Assay Percent Range 99%
    Molecular Formula ClLiO4
    Appearance White crystals
    MDL Number MFCD00011079
    CAS 7791-03-9
    Health Hazard 1 H272-H301-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Lithium perchlorate, anhydrous

    H272-H301-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit