Alfa Aesar™ Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO4

Manufacturer: Alfa Aesar™ 4592336

Quantity 500g

 View more versions of this product

Catalog No. AA4592336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13453-78-6
    Molecular Formula ClLiO4·3H2O
    Linear Formula LiClO4·3H2O
    Formula Weight 160.45 (106.40 Anhydrous)
    MDL Number MFCD00149765
    UN Number UN1481
    Merck Index 14,5539
    Chemical Name or Material Lithium perchlorate trihydrate
    Grade ACS Grade

    Specifications

    Formula Weight 160.45 (106.40 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13453-78-6
    Merck Index 14,5539
    TSCA TSCA
    Odor Odorless
    Linear Formula LiClO4·3H2O
    Color White
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 1.84g/mL
    MDL Number MFCD00149765
    UN Number UN1481
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
    RTECS Number SC8750000
    Physical Form Crystalline
    Molecular Formula ClLiO4·3H2O
    Health Hazard 1 Danger
    Concentration 63.0 to 68.0% LiClO4
    Chemical Name or Material Lithium perchlorate trihydrate
    Grade ACS Grade

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit