Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO4

Manufacturer:  Alfa Aesar™ 4592309

Quantity 10g

 View more versions of this product

Catalog No. AA4592309


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13453-78-6
LiClO4·3H2O
MFCD00149765
14,5539
ACS Grade
ClLiO4·3H2O
160.45 (106.40 Anhydrous)
UN1481
Lithium perchlorate trihydrate

Specifications

13453-78-6
ClLiO4·3H2O
160.45 (106.40 Anhydrous)
UN1481
63.0 to 68.0% LiClO4
Odorless
10g
ACS Grade
Crystalline
LiClO4·3H2O
MFCD00149765
14,5539
1.84g/mL
White
Lithium perchlorate trihydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

RTECSNumber : SC8750000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit