Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium peroxide, 95%, Acros Organics

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
12031-80-0
Li2O2
45.88
MFCD00016184
HPGPEWYJWRWDTP-UHFFFAOYSA-N
dilithium peroxide,lithium peroxide li2 o2,lithium oxide,unii-9anx556r5f,lithium peroxide un1472 oxidizer
25489
dilithium;peroxide
[Li+].[Li+].[O-][O-]
12031-80-0
Li2O2
45.88
MFCD00016184
HPGPEWYJWRWDTP-UHFFFAOYSA-N
dilithium peroxide,lithium peroxide li2 o2,lithium oxide,unii-9anx556r5f,lithium peroxide un1472 oxidizer
25489
dilithium;peroxide
[Li+].[Li+].[O-][O-]
Assay Percent Range 95%
Linear Formula LiO2Li
Formula Weight 45.88
Physical Form Powder
Percent Purity 95%
Packaging Plastic bottle
Color White to Yellow
Quantity 250g
Chemical Name or Material Lithium peroxide

GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 234-758-