Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium phenoxide, 0.6M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass Bottle
555-24-8,109-99-9
C6H5LiO
100.05
XAVQZBGEXVFCJI-UHFFFAOYSA-M
lithium phenoxide,lithium phenolate,phenol lithium salt,phenoxylithium,lithium phenoxide solution,lithium 1+ phenolate,lithium 1+ ion phenolate,lithium, 3-hydroxyphenyl-, lithium salt 9ci
11815295
lithium;phenoxide
[Li+].C1=CC=C(C=C1)[O-]
555-24-8,109-99-9
C6H5LiO
100.05
XAVQZBGEXVFCJI-UHFFFAOYSA-M
lithium phenoxide,lithium phenolate,phenol lithium salt,phenoxylithium,lithium phenoxide solution,lithium 1+ phenolate,lithium 1+ ion phenolate,lithium, 3-hydroxyphenyl-, lithium salt 9ci
11815295
lithium;phenoxide
[Li+].C1=CC=C(C=C1)[O-]
Formula Weight 100.05
Physical Form Solution
Concentration 0.55 to 0.65M
Density 0.9180g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.918
Boiling Point 67.0°C
Color Orange to Yellow
Quantity 100mL
Flash Point −19°C
Chemical Name or Material Lithium phenoxide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapour.
May form explosive peroxides.
Suspected of causing cancer.
Harmful if swallowed.
May

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 209-086-6