Alfa Aesar™ Lithium rod, 12.7mm (0.5 in.) dia. x approximately 165mm (6.50 in.) long, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 10773P2

Quantity 1 lb.

 View more versions of this product

Catalog No. AA10773P2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-93-2
    MDL Number MFCD00134051
    UN Number UN1415
    Chemical Name or Material Lithium rod, 12.7mm (0.5 in.) dia. x ≈165mm (6.50 in.) long, Mineral oil coated in vacuum can

    Specifications

    Health Hazard 3 GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H260-H314
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Color Silver-gray
    Weight ≈9 rods/0.25lb
    TSCA TSCA
    MDL Number MFCD00134051
    CAS 7439-93-2
    UN Number UN1415
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM
    Physical Form Rod
    RTECS Number OJ5540000
    Quantity 1 lb.
    Chemical Name or Material Lithium rod, 12.7mm (0.5 in.) dia. x ≈165mm (6.50 in.) long, Mineral oil coated in vacuum can
    Assay 99.9%, (metals basis)

    GHS H Statement
    H260-H314
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit