Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium sulfate, for analysis, anhydrous, Acros Organics

Click to view available options
Quantity:
250g
1kg
10377-48-7
Li2O4S
109.94
INHCSSUBVCNVSK-UHFFFAOYSA-L
lithium sulfate,lithium sulphate,dilithium sulfate,sulfuric acid, dilithium salt,lithiophor,li2so4,dilthium sulfate,sulfuric acid, lithium salt 1:2,unii-919xa137jk,lithium sulfate, anhydrous
66320
CHEBI:53474
dilithium;sulfate
[Li+].[Li+].[O-]S(=O)(=O)[O-]
10377-48-7
Li2O4S
109.94
INHCSSUBVCNVSK-UHFFFAOYSA-L
lithium sulfate,lithium sulphate,dilithium sulfate,sulfuric acid, dilithium salt,lithiophor,li2so4,dilthium sulfate,sulfuric acid, lithium salt 1:2,unii-919xa137jk,lithium sulfate, anhydrous
66320
CHEBI:53474
dilithium;sulfate
[Li+].[Li+].[O-]S(=O)(=O)[O-]
Linear Formula Li2SO4
Merck Index 15,5597
Solubility Information Solubility in water: 25.7 g/100mL (25°C). Other solubilities: practically insoluble in alcohol
Formula Weight 109.94
Grade Analysis
Calcium (Ca) 0.02% max.
Density 2.2200g/mL
Heavy Metals (as Pb) 0.002% max.
Iron (Fe) 0.005% max.
Magnesium (Mg) 0.02% max.
Potassium (K) 0.02% max.
Solubility (10% in water) Clear colorless to slightly hazy
Specific Gravity 2.22
Water 1% max.
Melting Point 859.0°C
Quantity 250g
Chemical Name or Material Lithium sulfate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Warning

EINECSNumber : 233-820-4