Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium sulfate monohydrate, 99+%, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
10102-25-7
Li2O4S·H2O
127.96
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
dilithium;sulfate;hydrate
[Li+].[Li+].O.[O-]S(=O)(=O)[O-]
10102-25-7
Li2O4S·H2O
127.96
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
dilithium;sulfate;hydrate
[Li+].[Li+].O.[O-]S(=O)(=O)[O-]
Assay Percent Range 99+%
Linear Formula Li2SO4·H2O
Merck Index 15,5597
Solubility Information Solubility in water: 295.5g/L (20°C)
Formula Weight 127.96
Percent Purity 99+%
Grade ACS Reagent
Chloride 0.002% max
Insoluble Matter 0.01% max
Packaging Plastic bottle
Solubility Solubility in water: 295.5g/L (20°C).
Melting Point 860.0°C
Quantity 500g
Chemical Name or Material Lithium sulfate monohydrate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pr

GHS Signal Word: Warning