Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium sulfate monohydrate, 99%, Acros Organics

Click to view available options
Quantity:
500g
10102-25-7
H2Li2O5S
127.951
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate,dilithium 1+ ion hydrate sulfate,dilithium sulfate hydrate,sufuric acid, dilithium slat, monohydrate,lithiumsulfatemonohydrate,dilithotab hydrate sulfate,lithium sulphate monohydrate,lithium sulfate, acs,ksc181s7n
165815
dilithium;sulfate;hydrate
[Li+].[Li+].O.[O-]S(=O)(=O)[O-]
10102-25-7
H2Li2O5S
127.951
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate,dilithium 1+ ion hydrate sulfate,dilithium sulfate hydrate,sufuric acid, dilithium slat, monohydrate,lithiumsulfatemonohydrate,dilithotab hydrate sulfate,lithium sulphate monohydrate,lithium sulfate, acs,ksc181s7n
165815
dilithium;sulfate;hydrate
[Li+].[Li+].O.[O-]S(=O)(=O)[O-]
Assay Percent Range 99%
Linear Formula Li2SO4·H2O
Formula Weight 127.96
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Solubility Solubility in water: 295.5g/L (20°C).
Melting Point 860.0°C
Quantity 500g
Chemical Name or Material Lithium sulfate monohydrate