Alfa Aesar™ Lithium tantalum ethoxide, 99.9% (metals basis), 5% w/v in ethanol

Precursor for Lithium tantalum oxide films

Manufacturer: Alfa Aesar™ 42344AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42344AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127503-04-2
    Linear Formula LiTa(OCH2CH3)6
    Formula Weight 458.04
    MDL Number MFCD00210630
    UN Number UN1170
    Chemical Name or Material Lithium tantalum ethoxide

    Specifications

    Formula Weight 458.04
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Health Hazard 1 Danger
    Color Pale yellow
    Odor Alcohol-like
    Sensitivity Moisture Sensitive
    Concentration 5% w/v in ethanol
    Linear Formula LiTa(OCH2CH3)6
    MDL Number MFCD00210630
    UN Number UN1170
    CAS 127503-04-2
    Quantity 100mL
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
    Chemical Name or Material Lithium tantalum ethoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit