Alfa Aesar™ Lithium tantalum isopropoxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4052009

Quantity 10g

 View more versions of this product

Catalog No. AA4052009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21864-24-4
    Linear Formula LiTa[OCH(CH3)2]6
    Formula Weight 542.42
    MDL Number MFCD00798538
    UN Number UN2925
    Chemical Name or Material Lithium tantalum isopropoxide

    Specifications

    Formula Weight 542.42
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Physical Form Wet Solid
    Quantity 10g
    UN Number UN2925
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H228
    Causes severe skin burns and eye damage.
    Flammable solid.
    MDL Number MFCD00798538
    Boiling Point 160° to 180°C (0.1mmHg)
    Linear Formula LiTa[OCH(CH3)2]6
    CAS 21864-24-4
    Color Off-white
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Sensitivity Moisture Sensitive
    Chemical Name or Material Lithium tantalum isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H228
    Causes severe skin burns and eye damage.
    Flammable solid.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit