Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium tantalum isopropoxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4052004

Quantity 2g

 View more versions of this product

Catalog No. AA4052004


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

21864-24-4
542.42
UN2925
LiTa[OCH(CH3)2]6
MFCD00798538
Lithium tantalum isopropoxide

Specifications

21864-24-4
160° to 180°C (0.1mmHg)
Moisture Sensitive
LiTa[OCH(CH3)2]6
MFCD00798538
Lithium tantalum isopropoxide
2g
Off-white
Wet Solid
542.42
UN2925
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit