Alfa Aesar™ Lithium tantalum oxide, congruent, Puratronic™, 99.998% (metals basis)

Manufacturer: Alfa Aesar™ 1074509

Quantity 10g

 View more versions of this product

Catalog No. AA1074509

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12031-66-2
    Formula Weight 235.89
    Synonym Lithium metatantalate
    Chemical Name or Material Lithium tantalum oxide, Congruent
    Grade Puratronic™

    Specifications

    Formula Weight 235.89
    CAS 12031-66-2
    TSCA TSCA
    Synonym Lithium metatantalate
    Odor Odorless
    Physical Form Powder
    Quantity 10g
    Chemical Name or Material Lithium tantalum oxide, Congruent
    Assay 99.998%, (metals basis)
    Grade Puratronic™

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit