Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tert-butoxide, 1M solution in hexanes, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
1907-33-1,110-54-3
C4H9LiO
80.055
MFCD00050479
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
1907-33-1,110-54-3
C4H9LiO
80.055
MFCD00050479
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
Appearance of Solution Clear yellow solution
Linear Formula LiOCH(CH3)3
Concentration 0.95 to 1.10M
Density 0.69g/mL
Formula Weight 80.06
Physical Form May Contain Some Turbidity or Precipitate
Packaging AcroSeal™ Glass Bottle
Quantity 800mL
Chemical Name or Material Lithium tert-butoxide, 1M solution in hexanes

GHS H Statement:
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of dama

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger