Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium tert-butoxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4413330

Quantity 250g

 View more versions of this product

Catalog No. AA4413330


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1907-33-1
(CH3)3COLi
MFCD00050479
3620018
C4H9LiO
80.06
UN3206
Lithium tert-butoxide

Specifications

≈150°C (dec.)
250g
Odorless
Powder
(CH3)3COLi
MFCD00050479
3620018
99.9%, (metals basis)
1907-33-1
White to cream
Air & Moisture Sensitive
C4H9LiO
80.06
UN3206
Lithium tert-butoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H251-H314-H302
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

RTECSNumber : UB8520000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit